Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZSCAN12
Entrez Gene ID
102171019
Gene Name
zinc finger and SCAN domain containing 12
Gene Aliases
-
Chromosome Location
Chr.23: 19585210 - 19594163 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018038516.1 XP_017894005.1 1 - 2 777 73

View Full Product Details