Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CAPS2
Entrez Gene ID
102179070
Gene Name
calcyphosine 2
Gene Aliases
-
Chromosome Location
Chr.5: 4589549 - 4642786 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013963641.2 XP_013819095.2 10 - 11 1029 93

View Full Product Details