Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SUOX
Entrez Gene ID
102174192
Gene Name
sulfite oxidase
Gene Aliases
-
Chromosome Location
Chr.5: 56689630 - 56694245 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018048094.1 XP_017903583.1 2 - 3 444 66
XM_018048095.1 XP_017903584.1 2 - 3 430 66
XM_018048096.1 XP_017903585.1 3 - 4 511 66
XM_018048097.1 XP_017903586.1 2 - 3 145 66
GenBank mRNA EU418447.2 - 2 - 3 111 66
JO429150.2 - 2 - 3 153 66
JO502728.2 - 2 - 3 166 66

View Full Product Details