Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC35C1
Entrez Gene ID
102190895
Gene Name
solute carrier family 35 member C1
Gene Aliases
-
Chromosome Location
Chr.15: 7132178 - 7140431 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018059194.1 XP_017914683.1 2 - 3 588 87
XM_018059195.1 XP_017914684.1 2 - 3 604 87
XM_018059196.1 XP_017914685.1 2 - 3 569 87
GenBank mRNA JO602989.2 - - 1175 87

View Full Product Details