Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CFHR5
Entrez Gene ID
102170257
Gene Name
complement factor H related 5
Gene Aliases
-
Chromosome Location
Chr.16: 74581542 - 74609412 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005691048.3 XP_005691105.2 5 - 6 882 83

View Full Product Details