Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM64
Entrez Gene ID
102172825
Gene Name
transmembrane protein 64
Gene Aliases
-
Chromosome Location
Chr.14: 8656708 - 8693107 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA JO402587.2 - 1 - 2 602 109
JO443824.2 - 1 - 2 197 109

View Full Product Details