Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLRF2
Entrez Gene ID
102189376
Gene Name
killer cell lectin like receptor F2
Gene Aliases
-
Chromosome Location
Chr.5: 98490905 - 98510035 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018049112.1 XP_017904601.1 4 - 5 362 82

View Full Product Details