Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM58
Entrez Gene ID
102190778
Gene Name
tripartite motif containing 58
Gene Aliases
-
Chromosome Location
Chr.7: 69184394 - 69197774 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005682840.2 XP_005682897.2 2 - 3 515 72
GenBank mRNA JO338343.2 - 4 - 5 397 72
JR121379.1 - - 270 72

View Full Product Details