Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FBXO30
Entrez Gene ID
102184435
Gene Name
F-box protein 30
Gene Aliases
-
Chromosome Location
Chr.9: 70139831 - 70157216 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018053307.1 XP_017908796.1 2 - 3 2249 67
GenBank mRNA JT650546.1 - 6 - 7 1122 67

View Full Product Details