Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- MOB2
- Entrez Gene ID
-
102187682
- Gene Name
- MOB kinase activator 2
- Gene Aliases
-
-
- Chromosome Location
-
Chr.29: 50987205 - 51039263 on Build ARS1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
JO612584.2 |
- |
- |
1268 |
91 |
- Gene Symbol
- ZC3H12D
- Entrez Gene ID
-
108633194
- Gene Name
- zinc finger CCCH-type containing 12D
- Gene Aliases
-
-
- Chromosome Location
-
Chr.9: 73643883 - 73678439 on Build ARS1
View Full Product Details