Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC45A3
Entrez Gene ID
102182991
Gene Name
solute carrier family 45 member 3
Gene Aliases
-
Chromosome Location
Chr.16: 2813492 - 2832220 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018060056.1 XP_017915545.1 5 - 6 1614 62
XM_018060057.1 XP_017915546.1 5 - 6 1678 62
XM_018060058.1 XP_017915547.1 4 - 5 1539 62
XM_018060059.1 XP_017915548.1 3 - 4 6057 62
XM_018060060.1 XP_017915549.1 4 - 5 1603 62
XM_018060061.1 XP_017915550.1 4 - 5 2463 62
GenBank mRNA KA315861.1 - 1 - 2 1769 62

View Full Product Details