Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TPRG1L
Entrez Gene ID
102183086
Gene Name
tumor protein p63 regulated 1 like
Gene Aliases
-
Chromosome Location
Chr.16: 47983830 - 47987523 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018060465.1 XP_017915954.1 4 - 5 759 66
XM_018060466.1 XP_017915955.1 4 - 5 760 66
GenBank mRNA JO432228.2 - 4 - 5 550 66

View Full Product Details