Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
MAGIX
Entrez Gene ID
102191122
Gene Name
MAGI family member, X-linked
Gene Aliases
-
Chromosome Location
Chr.Un|NW_017190157: 111924 - 113527 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018045144.1 XP_017900633.1 - 844 83

View Full Product Details