Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ASTL
Entrez Gene ID
102173096
Gene Name
astacin like metalloendopeptidase
Gene Aliases
-
Chromosome Location
Chr.11: 2199482 - 2215579 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018054700.1 XP_017910189.1 4 - 5 378 89

View Full Product Details