Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PARD6A
Entrez Gene ID
102185594
Gene Name
par-6 family cell polarity regulator alpha
Gene Aliases
-
Chromosome Location
Chr.18: 36475122 - 36477289 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005692148.2 XP_005692205.1 2 - 3 664 74

View Full Product Details