Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLEC4F
Entrez Gene ID
102185659
Gene Name
C-type lectin domain family 4 member F
Gene Aliases
-
Chromosome Location
Chr.11: 13451331 - 13463639 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005686384.3 XP_005686441.1 4 - 5 1549 67
XM_018055079.1 XP_017910568.1 4 - 5 1787 67

View Full Product Details