Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LCN6
Entrez Gene ID
102186775
Gene Name
lipocalin 6
Gene Aliases
-
Chromosome Location
Chr.11: 104609030 - 104613951 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018054747.1 XP_017910236.1 1 - 2 89 64

View Full Product Details