Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RESP18
Entrez Gene ID
102179426
Gene Name
regulated endocrine specific protein 18
Gene Aliases
-
Chromosome Location
Chr.2: 28537447 - 28542997 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005676587.3 XP_005676644.1 3 - 4 443 87
GenBank mRNA JO645882.2 - 3 - 4 439 87

View Full Product Details