Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C15H11orf74
Entrez Gene ID
102173404
Gene Name
chromosome 15 C11orf74 homolog
Gene Aliases
-
Chromosome Location
Chr.15: 15839297 - 15908950 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005690093.2 XP_005690150.2 4 - 5 503 122
XM_005690094.2 XP_005690151.2 5 - 6 561 122
XM_013969830.2 XP_013825284.2 3 - 4 347 122
GenBank mRNA KA326007.1 - 4 - 5 493 122

View Full Product Details