Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LRRC30
Entrez Gene ID
102172955
Gene Name
leucine rich repeat containing 30
Gene Aliases
-
Chromosome Location
Chr.24: 40488615 - 40489770 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005697127.3 XP_005697184.2 1 - 1 662 73

View Full Product Details