Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TMEM268
Entrez Gene ID
102177629
Gene Name
transmembrane protein 268
Gene Aliases
-
Chromosome Location
Chr.8: 103502709 - 103531090 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018052670.1 XP_017908159.1 6 - 7 748 67
XM_018052671.1 XP_017908160.1 6 - 7 748 67
XM_018052672.1 XP_017908161.1 5 - 6 661 67
XM_018052673.1 XP_017908162.1 6 - 7 748 67
XM_018052674.1 XP_017908163.1 6 - 7 748 67
GenBank mRNA KA314190.1 - 6 - 7 713 67

View Full Product Details