Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NLRP8
Entrez Gene ID
102170455
Gene Name
NLR family pyrin domain containing 8
Gene Aliases
-
Chromosome Location
Chr.18: 64978163 - 64993714 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018063248.1 XP_017918737.1 11 - 12 3384 66

View Full Product Details