Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SH2D6
Entrez Gene ID
102187235
Gene Name
SH2 domain containing 6
Gene Aliases
-
Chromosome Location
Chr.11: 49079743 - 49087049 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018055381.1 XP_017910870.1 13 - 14 755 77

View Full Product Details