Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC102189642
Entrez Gene ID
102189642
Gene Name
uncharacterized LOC102189642
Gene Aliases
OFCC1
Chromosome Location
Chr.23: 4117949 - 4365730 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018038501.1 XP_017893990.1 7 - 8 798 99

View Full Product Details