Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TXNIP
Entrez Gene ID
102183379
Gene Name
thioredoxin interacting protein
Gene Aliases
-
Chromosome Location
Chr.3: 99143349 - 99147525 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005677732.3 XP_005677789.1 5 - 6 1098 104

View Full Product Details