Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPRED3
Entrez Gene ID
102176992
Gene Name
sprouty related EVH1 domain containing 3
Gene Aliases
-
Chromosome Location
Chr.18: 49097377 - 49106412 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018062336.1 XP_017917825.1 3 - 4 715 90

View Full Product Details