Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C9orf72
Entrez Gene ID
102168257
Gene Name
C9orf72-SMCR8 complex subunit
Gene Aliases
C8H9orf72
Chromosome Location
Chr.8: 16647898 - 16674101 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018051885.1 XP_017907374.1 4 - 5 717 118
GenBank mRNA KA329469.1 - 4 - 5 679 118

View Full Product Details