Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRMT12
Entrez Gene ID
102189221
Gene Name
tRNA methyltransferase 12 homolog
Gene Aliases
-
Chromosome Location
Chr.14: 66408335 - 66414451 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005688858.3 XP_005688915.2 1 - 2 1556 94

View Full Product Details