Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLAC8L1
Entrez Gene ID
102169001
Gene Name
PLAC8 like 1
Gene Aliases
-
Chromosome Location
Chr.7: 53136236 - 53159276 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005683098.3 XP_005683155.1 2 - 3 317 78

View Full Product Details