Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TAFA1
Entrez Gene ID
102177386
Gene Name
TAFA chemokine like family member 1
Gene Aliases
FAM19A1
Chromosome Location
Chr.22: 32905060 - 33423807 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005695754.2 XP_005695811.1 3 - 4 750 62

View Full Product Details