Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
NDUFA11
Entrez Gene ID
102181026
Gene Name
NADH:ubiquinone oxidoreductase subunit A11
Gene Aliases
-
Chromosome Location
Chr.7: 92032030 - 92037341 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005682506.3 XP_005682563.1 2 - 3 481 61

View Full Product Details