Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PIM2
Entrez Gene ID
102170766
Gene Name
Pim-2 proto-oncogene, serine/threonine kinase
Gene Aliases
-
Chromosome Location
Chr.Un|NW_017190136: 97060 - 103902 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005700764.3 XP_005700821.2 1 - 2 1777 69
GenBank mRNA JT705205.1 - 1 - 2 442 69

View Full Product Details