Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
KLF4
Entrez Gene ID
102171875
Gene Name
Kruppel like factor 4
Gene Aliases
-
Chromosome Location
Chr.8: 96960669 - 96965428 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA KA329193.1 - - 1136 79
KU041754.1 - 3 - 4 1163 79

View Full Product Details