Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KLK4
Entrez Gene ID
102177461
Gene Name
kallikrein related peptidase 4
Gene Aliases
-
Chromosome Location
Chr.18: 58002484 - 58006407 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018062888.1 XP_017918377.1 4 - 5 613 70

View Full Product Details