Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CENPX
Entrez Gene ID
102178856
Gene Name
centromere protein X
Gene Aliases
STRA13
Chromosome Location
Chr.19: 50261728 - 50265357 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018065436.1 XP_017920925.1 3 - 4 184 106

View Full Product Details