Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GIMAP8
Entrez Gene ID
102191585
Gene Name
GTPase, IMAP family member 8
Gene Aliases
-
Chromosome Location
Chr.4: 7783430 - 7829255 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018047475.1 XP_017902964.1 3 - 4 909 100

View Full Product Details