Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CTDSP2
Entrez Gene ID
102168891
Gene Name
CTD small phosphatase 2
Gene Aliases
-
Chromosome Location
Chr.5: 54975053 - 54998110 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005680257.3 XP_005680314.1 6 - 7 1031 79

View Full Product Details