Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DUSP5
Entrez Gene ID
102186181
Gene Name
dual specificity phosphatase 5
Gene Aliases
-
Chromosome Location
Chr.26: 20923765 - 20936379 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018041694.1 XP_017897183.1 2 - 3 714 83

View Full Product Details