Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C13H20orf27
Entrez Gene ID
102180506
Gene Name
chromosome 13 C20orf27 homolog
Gene Aliases
-
Chromosome Location
Chr.13: 50785602 - 50799452 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018057399.1 XP_017912888.1 5 - 6 1406 73
GenBank mRNA JO413267.2 - 1 - 2 642 73

View Full Product Details