Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MPPED1
Entrez Gene ID
102175011
Gene Name
metallophosphoesterase domain containing 1
Gene Aliases
-
Chromosome Location
Chr.5: 113138117 - 113221869 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018048878.1 XP_017904367.1 3 - 4 657 68

View Full Product Details