Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZMYM5
Entrez Gene ID
102172456
Gene Name
zinc finger MYM-type containing 5
Gene Aliases
-
Chromosome Location
Chr.12: 50494767 - 50519959 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018056524.1 XP_017912013.1 7 - 8 1430 106
XM_018056525.1 XP_017912014.1 7 - 8 1450 106
XM_018056526.1 XP_017912015.1 7 - 8 1562 106

View Full Product Details