Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KRT39
Entrez Gene ID
102178207
Gene Name
keratin 39
Gene Aliases
-
Chromosome Location
Chr.19: 40797168 - 40815645 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA KA326566.1 - 1 - 2 2322 61

View Full Product Details