Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C19H17orf64
Entrez Gene ID
102191645
Gene Name
chromosome 19 C17orf64 homolog
Gene Aliases
-
Chromosome Location
Chr.19: 12166508 - 12178018 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018064063.1 XP_017919552.1 3 - 4 329 104

View Full Product Details