Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Crybb3
Entrez Gene ID
100306963
Gene Name
crystallin beta B3
Gene Aliases
-
Chromosome Location
Chr.Un|NT_176415: 17357587 - 17385716 on Build Cavpor3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001173070.1 NP_001166541.1 3 - 4 267 59
GenBank mRNA EB370163.1 - 3 - 4 254 59
EF457997.1 - 3 - 4 267 59

View Full Product Details