Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DnaJ-H
Entrez Gene ID
34707
Gene Name
DnaJ homolog
Gene Aliases
CG9828, DNA J-H, DNAJ-H, Dmel\CG9828, Dmel_CG9828, NEST:bs08d07, dDnaJ-H, dHdj2, dnaJ-H
Chromosome Location
Chr.2L: 13163209 - 13166072 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0032474
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273502.1 NP_001260431.1 1 - 2 198 69
NM_135761.3 NP_609605.1 1 - 2 198 69
NM_165025.2 NP_723785.1 2 - 3 700 69
GenBank mRNA BT150251.1 - 2 - 3 700 69

View Full Product Details