Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Chro
Entrez Gene ID
40508
Gene Name
Chromator
Gene Aliases
CG10712, CHRIZ, CHRIZ/CHRO, CHRO, CHRO/CHRIZ, Chiz, Chr, Chriz, Chro(Chriz), Dmel\CG10712, Dmel_CG10712, MRG15, anon-EST:fe2E3, chro
Chromosome Location
Chr.3L: 22873702 - 22878984 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0044324
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001275297.1 NP_001262226.1 3 - 4 295 64
NM_001300221.1 NP_001287150.1 2 - 3 436 64
NM_141169.3 NP_649426.3 1 - 2 251 64
NM_168975.2 NP_730762.2 2 - 3 220 64
NM_168976.2 NP_730763.2 2 - 3 151 64
GenBank mRNA BT128821.1 - 2 - 3 190 64

View Full Product Details