Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MED8
- Entrez Gene ID
-
37305
- Gene Name
- Mediator complex subunit 8
- Gene Aliases
-
ARC32, Arc32, CG13867, CG30140, Dmel\CG13867, Dmel_CG13867, Med8, Med8/ARC32, dARC32
- Chromosome Location
-
Chr.2R: 20325844 - 20326806 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0034503
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_153772.2 |
NP_722456.1 |
1 - 1 |
626 |
75 |
GenBank mRNA |
BT050506.1 |
- |
1 - 1 |
626 |
75 |
View Full Product Details