Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cpr49Aa
Entrez Gene ID
246413
Gene Name
Cuticular protein 49Aa
Gene Aliases
CG30045, CG8496, DmelCpr49Aa, Dmel\CG30045, Dmel_CG30045
Chromosome Location
Chr.2R: 12376626 - 12377378 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0050045
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103815.3 NP_001097285.1 2 - 3 292 65

View Full Product Details