Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ftz-f1
Entrez Gene ID
40045
Gene Name
ftz transcription factor 1
Gene Aliases
BFTZ-F1, CG13378, CG4059, DEP1, DmFTZ-F1, Dmel\CG4059, Dmel_CG4059, FIP1, FTZ-F1, FTZ-F1A, FTZ-F1alpha, FTZ-Fl, FTZF1, FTZf1alpha, Ftz-F1, Ftz-f1, Ftzf1, NR5A3, alphaFTZ-F1, alphaFTZF1, alphaftz-f1, bFTZ-F1, beta FTZ-F1, beta Ftz-f1, beta-FTZ-F1, beta-Ftz-F1, betaFTZ-F1, betaFTZF1, betaFtz-F1, betaFtz-f1, betaftz-F1, betaftz-f1, betaftzf1, dFTZ-F1alpha, dFtz-F1alpha, dmFTZ-f1, ftz-F1, ftz-f1alpha, ftzf1, l(3)03649
Chromosome Location
Chr.3L: 18750607 - 18801309 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0001078
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001259895.2 NP_001246824.1 2 - 3 2640 90
NM_001316437.1 NP_001303366.1 2 - 3 2640 90
NM_079419.3 NP_524143.2 4 - 5 3279 90
NM_168775.2 NP_730359.1 2 - 3 1852 90
GenBank mRNA BT058016.1 - 4 - 5 2865 90

View Full Product Details