Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ir76a
Entrez Gene ID
40157
Gene Name
Ionotropic receptor 76a
Gene Aliases
CG34257, CG42584, CG8533, CT24909, DmelIR76a, Dmel\CG42584, Dmel_CG42584, IR76a, ir76a
Chromosome Location
Chr.3L: 19799278 - 19803427 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0260874
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104177.3 NP_001097647.3 5 - 6 845 74
NM_001144510.3 NP_001137982.1 3 - 4 589 74
NM_140891.4 NP_649148.2 3 - 4 576 74
NM_168817.3 NP_730452.2 4 - 5 615 74

View Full Product Details